ENA429361180 Enamine

5-[(2,4-dichlorophenyl)methoxy]-1,3,4-thiadiazol-2-amine

  • MDL number MFCD18446992
  • Cas Number : 1269527-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191924

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191924
Availability details for
Quantity
  • Availability
  • Add to Cart