ENA429361187 Enamine

1-bromo-3-fluorobutane

  • MDL number MFCD20621878

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191949

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191949
Availability details for
Quantity
  • Availability
  • Add to Cart