ENA429361188 Enamine

N-ethylazetidin-3-amine dihydrochloride

  • MDL number MFCD10565783
  • Cas Number : 149088-17-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191950

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191950
Availability details for
Quantity
  • Availability
  • Add to Cart