ENA429361191 Enamine

7-{[(2-aminoethyl)sulfanyl]methyl}-5H-[1,3,4]thiadiazolo[3,2-a]pyrimidin-5-one dihydrochloride

  • MDL number MFCD17430415
  • Cas Number : 1245569-87-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191954

Safety Information

msds https://www.enaminestore.com/catalog/EN300-191954
Availability details for
Quantity
  • Availability
  • Add to Cart