ENA429361194 Enamine

3-chloro-4-[(dimethylamino)methyl]aniline

  • MDL number MFCD10021509
  • Cas Number : 879222-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-191958

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191958
Availability details for
Quantity
  • Availability
  • Add to Cart