ENA429361195 Enamine

2-(pyridin-3-yl)cyclopentan-1-ol

  • MDL number MFCD16818413
  • Cas Number : 1247593-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191964

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191964
Availability details for
Quantity
  • Availability
  • Add to Cart