ENA429361196 Enamine

2-(methylsulfanyl)cyclopentane-1-carboxylic acid

  • MDL number MFCD17288066
  • Cas Number : 1339208-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-191988

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-191988
Availability details for
Quantity
  • Availability
  • Add to Cart