ENA429361202 Enamine

1-cyclopropylpyrrolidine-3-carbonitrile

  • MDL number MFCD17263683
  • Cas Number : 1341409-48-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189822

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189822
Availability details for
Quantity
  • Availability
  • Add to Cart