ENA429361203 Enamine

prop-2-en-1-yl 4-hydroxypiperidine-1-carboxylate

  • MDL number MFCD08694514
  • Cas Number : 187265-40-3
  • Molecular Weight : 185.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -10
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-189823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189823
Availability details for
Quantity
  • Availability
  • Add to Cart