ENA429361218 Enamine

methyl 3,3-difluorocyclopentane-1-carboxylate

  • MDL number MFCD22572330
  • Cas Number : 1394129-94-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192032

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192032
Availability details for
Quantity
  • Availability
  • Add to Cart