ENA429361220 Enamine

2-(2-hydroxyphenyl)-2-oxoethane-1-sulfonamide

  • MDL number MFCD24718626
  • Cas Number : 74538-97-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192036

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192036
Availability details for
Quantity
  • Availability
  • Add to Cart