ENA429361221 Enamine

3,4-dichloro-2,6-dinitrophenol

  • MDL number MFCD09836174
  • Cas Number : 1664-10-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192038

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192038
Availability details for
Quantity
  • Availability
  • Add to Cart