ENA429361222 Enamine

3-(4-aminophenyl)imidazolidine-2,4-dione

  • MDL number MFCD09811313
  • Cas Number : 625094-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192039
Availability details for
Quantity
  • Availability
  • Add to Cart