ENA429361225 Enamine

3-(4-iodo-3,5-dimethyl-1H-pyrazol-1-yl)-1lambda6-thiolane-1,1-dione

  • MDL number MFCD22731852
  • Cas Number : 1536708-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189827
Availability details for
Quantity
  • Availability
  • Add to Cart