ENA429361227 Enamine

2,2-difluoro-1-[3-(propan-2-yloxy)phenyl]ethan-1-one

  • MDL number MFCD26045131
  • Cas Number : 1565091-61-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 48-50
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192051

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192051
Availability details for
Quantity
  • Availability
  • Add to Cart