ENA429361229 Enamine

ethyl 2-(1,1-dioxo-1$l^{6}-thietan-3-ylidene)acetate

  • MDL number MFCD22421708
  • Cas Number : 1394319-79-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192053

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192053
Availability details for
Quantity
  • Availability
  • Add to Cart