ENA429361233 Enamine

2,5-dioxo-2,5-dihydrofuran-3-sulfonyl chloride

  • MDL number MFCD19200173
  • Cas Number : 40336-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192058

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192058
Availability details for
Quantity
  • Availability
  • Add to Cart