ENA429361237 Enamine

2-(1-methyl-1H-pyrazol-4-yl)pyrimidine-5-carbaldehyde

  • MDL number MFCD21131866
  • Cas Number : 1484524-86-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192092
Availability details for
Quantity
  • Availability
  • Add to Cart