ENA429361238 Enamine

3-bromo-4-phenoxybenzene-1-sulfonyl chloride

  • MDL number MFCD16512196
  • Cas Number : 1284960-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192103

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192103
Availability details for
Quantity
  • Availability
  • Add to Cart