ENA429361240 Enamine

tert-butyl N-[3-hydroxy-2-(2-methylpropyl)propyl]carbamate

  • MDL number MFCD18433206
  • Cas Number : 1692710-67-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192131

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192131
Availability details for
Quantity
  • Availability
  • Add to Cart