ENA429361242 Enamine

5-{[(tert-butoxy)carbonyl]amino}-2-methylbenzoic acid

  • MDL number MFCD17170598
  • Cas Number : 143617-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192133

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-192133
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart