ENA429361244 Enamine

3-methanesulfonamido-5-(trifluoromethyl)benzoic acid

  • MDL number MFCD13293235
  • Cas Number : 1291361-71-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189849

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189849
Availability details for
Quantity
  • Availability
  • Add to Cart