ENA429361250 Enamine

4-chloro-2,5-dimethylbenzoic acid

  • MDL number MFCD11845877
  • Cas Number : 90649-77-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192155

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192155
Availability details for
Quantity
  • Availability
  • Add to Cart