ENA429361251 Enamine

4-(1-cyano-1-methylethyl)benzoic acid

  • MDL number MFCD12761251
  • Cas Number : 129488-74-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192157

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192157
Availability details for
Quantity
  • Availability
  • Add to Cart