ENA429361256 Enamine

2-difluoromethanesulfonylbenzene-1-sulfonyl chloride

  • MDL number MFCD09738489
  • Cas Number : 63647-67-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192165

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192165
Availability details for
Quantity
  • Availability
  • Add to Cart