ENA429361257 Enamine

3-(4-chlorophenyl)butan-2-one

  • MDL number MFCD21775886
  • Cas Number : 21905-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192167
Availability details for
Quantity
  • Availability
  • Add to Cart