ENA429361259 Enamine

4-benzoylpyrimidine

  • MDL number MFCD13193451
  • Cas Number : 68027-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192171

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192171
Availability details for
Quantity
  • Availability
  • Add to Cart