ENA429361260 Enamine

4-(pyridine-3-carbonyl)pyrimidine

  • MDL number MFCD16988796
  • Cas Number : 188630-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192172

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192172
Availability details for
Quantity
  • Availability
  • Add to Cart