ENA429361266 Enamine

methyl 3-[2-oxo-5-(trifluoromethyl)-1,2-dihydropyridin-1-yl]propanoate

  • MDL number MFCD19444739
  • Cas Number : 1281936-66-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192207

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192207
Availability details for
Quantity
  • Availability
  • Add to Cart