ENA429361267 Enamine

(5S)-9-fluoro-2,3,4,5-tetrahydro-1-benzoxepin-5-ol

  • MDL number MFCD21295160
  • Cas Number : 1568193-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192225
Availability details for
Quantity
  • Availability
  • Add to Cart