ENA429361274 Enamine

2-(quinolin-3-yl)propanoic acid

  • MDL number MFCD16748884
  • Cas Number : 1247842-46-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192234
Availability details for
Quantity
  • Availability
  • Add to Cart