ENA429361276 Enamine

4-[5-(difluoromethyl)-1,2,4-oxadiazol-3-yl]benzoic acid

  • MDL number MFCD27261574
  • Cas Number : 1788846-47-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192237

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192237
Availability details for
Quantity
  • Availability
  • Add to Cart