ENA429361277 Enamine

5-[(morpholin-4-yl)methyl]pyridine-2-carboxylic acid

  • MDL number MFCD18261502
  • Cas Number : 1211591-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192238
Availability details for
Quantity
  • Availability
  • Add to Cart