ENA429361283 Enamine

3-bromo-4,5-dimethyl-4H-1,2,4-triazole

  • MDL number MFCD26679013
  • Cas Number : 56616-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192265

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192265
Availability details for
Quantity
  • Availability
  • Add to Cart