ENA429361290 Enamine

1-[4-bromo-2-(trifluoromethyl)phenyl]propan-1-one

  • MDL number MFCD16841977
  • Cas Number : 1251389-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192278

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192278
Availability details for
Quantity
  • Availability
  • Add to Cart