ENA429361296 Enamine

5-formyl-2,4-dimethylbenzoic acid

  • MDL number MFCD20651233
  • Cas Number : 1379174-31-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177-178
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192291

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192291
Availability details for
Quantity
  • Availability
  • Add to Cart