ENA429361298 Enamine

7'-chloro-3',4'-dihydro-1'H-spiro[oxolane-3,2'-quinoxaline]-3'-one

  • MDL number MFCD24215280
  • Cas Number : 1547735-92-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192299

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192299
Availability details for
Quantity
  • Availability
  • Add to Cart