ENA429361301 Enamine

2-amino-3-methyl-2-phenylbutanoic acid

  • MDL number MFCD00037751
  • Cas Number : 188586-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192326

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192326
Availability details for
Quantity
  • Availability
  • Add to Cart