ENA429361305 Enamine

2-hydroxy-3H-imidazo[4,5-b]pyridine-7-carboxylic acid

  • MDL number MFCD26047561
  • Cas Number : 1565404-86-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-189901

Safety Information

msds https://www.enaminestore.com/catalog/EN300-189901
Availability details for
Quantity
  • Availability
  • Add to Cart