ENA429361307 Enamine

2-(1H-indazol-5-yl)acetic acid

  • MDL number MFCD09925211
  • Cas Number : 933694-85-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192370

Safety Information

msds https://www.enaminestore.com/catalog/EN300-192370
Availability details for
Quantity
  • Availability
  • Add to Cart