ENA429361308 Enamine

methyl N-[3-(hydroxymethyl)phenyl]carbamate

  • MDL number MFCD12777042
  • Cas Number : 81863-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192376
Availability details for
Quantity
  • Availability
  • Add to Cart