ENA429361309 Enamine

2-trifluoromethanesulfonylbenzene-1-thiol

  • MDL number MFCD12174235
  • Cas Number : 1156000-34-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-192377

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192377
Availability details for
Quantity
  • Availability
  • Add to Cart