ENA429361312 Enamine

4-(1H-indol-3-yl)butanamide

  • MDL number MFCD00458388
  • Cas Number : 6245-91-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-189902

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189902
Availability details for
Quantity
  • Availability
  • Add to Cart