ENA429361373 Enamine

4-[(3-methoxyphenyl)methyl]piperidin-4-ol hydrochloride

  • MDL number MFCD28125245
  • Cas Number : 1788624-49-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192430

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192430
Availability details for
Quantity
  • Availability
  • Add to Cart