ENA429361387 Enamine

2-(1-methyl-1H-imidazol-5-yl)oxan-4-amine dihydrochloride

  • MDL number MFCD28125261
  • Cas Number : 1786220-99-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192613
Availability details for
Quantity
  • Availability
  • Add to Cart