ENA429361391 Enamine

bis[2-(1,6-dimethyl-1H-indol-3-yl)ethyl]azanium chloride

  • Cas Number : 2080412-75-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data Cl-
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192733

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192733
Availability details for
Quantity
  • Availability
  • Add to Cart