ENA429361394 Enamine

tert-butyl N-{3-hydroxy-2-[(pyridin-4-yl)methyl]propyl}carbamate

  • MDL number MFCD28125271
  • Cas Number : 1803562-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192751
Availability details for
Quantity
  • Availability
  • Add to Cart