ENA429361402 Enamine

(2R,3R)-2-(3-chlorophenyl)-1-methyl-5-oxopyrrolidine-3-carboxamide

  • MDL number MFCD28656990
  • Cas Number : 1820571-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-192810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192810
Availability details for
Quantity
  • Availability
  • Add to Cart