ENA429361406 Enamine

(2R,3R)-2-(1,5-dimethyl-1H-pyrazol-4-yl)-1-methyl-5-oxopyrrolidine-3-carboxamide

  • MDL number MFCD28657138
  • Cas Number : 1820569-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-192827

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192827
Availability details for
Quantity
  • Availability
  • Add to Cart