ENA429361408 Enamine

2-bromo-4-methyl-5-(prop-2-yn-1-yloxy)pyridine

  • MDL number MFCD28125285
  • Cas Number : 1788530-86-8
  • Molecular Weight : 226.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-192876

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-192876
Availability details for
Quantity
  • Availability
  • Add to Cart